Coplin Staining Jar, Globe Scientific Inc.
Supplier: Globe Scientific
513200
76170-006CS 117.22 USD
76170-006
Coplin Staining Jar, Globe Scientific Inc.
Jars Staining Jars
Made from durable, autoclavable polypropylene.
  • Large opening for easy placement and removal of slides
  • Screw-on cap prevents evaporation and contamination
  • Five slots hold up to ten slides back-to-back for staining