Moticam® 3+ Microscope Camera, Motic Instruments
Supplier: Motic
1100600100791
75784-746EA 876 USD
75784-746
Moticam® 3+ Microscope Camera, Motic Instruments
Cameras Digital Cameras
The Moticam® 3+ is a highly capable 3 megapixel microscope camera, perfect for use in universities, clinics and veterinary centers. Its USB 3 connection provides rapid data transfer speeds, resulting in impressive frame rates, and it comes packed with a multitude of accessories so you are ready to take microscope pictures the moment you open the box. All this and more makes the Moticam® 3+ a great value for any microscope user.
The Moticam® 3+ offers a 3 megapixel resolution, perfect for routine microscope imaging
The lastest version of Motic Images Plus software is included, giving users the ability to not only view their images, but also measure, annotate, report and adjust them as well. No license fees or upgrade fees necessary. Just register your Moticam® online and receive free updates as they are made available.
The Moticam® 3+ comes with everything you need to get started right away. The full package includes the camera, a focusable coated glass lens, two adjustable size eyepiece adapters, a 4-dot calibration slide, a macro viewing tube, all necessary cables, a standard tripod mount and a full version of Motic Images Plus software.
Its USB 3 connection makes for a lightining fast framerate, and its included software and accessories add up to an impressive and affordable package, ideal for universities and clinics.