Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plum Duo Eye Wash Bottles/Station Refills, Sterile Saline and Sterile pH Neutralizing, SP Scienceware

Supplier: Bel-Art Products
F24880-8103 F24880-8023
Uniquely designed Plum Duo eye wash allows rinsing of both eyes at the same time, eliminating the need to choose which eye to rinse first in an emergency

  • Dual eye piece keeps eyes open allowing delivery of solution to both eyes at the same time
  • Two gentle streams of solution flow from the inner eyes toward the outside to keep tear ducts free from further contamination
  • Dispense upright or inverted; quick twist caps break seal for immediate access
  • pH Neutralizing solution neutralizes acids and alkalis to a harmless level in 20 seconds or less
  • 0.9% sterile saline mimics human tears for effective rinsing of dirt and debris
  • Pictogram instructions overcome language barriers
  • Use as station refills or in mobile/portable first aid kits
  • Rinse times: pH Neutralizing – approximately 2 min./bottle, Saline – approximately 5 min./bottle
  • No maintenance bottles; 3-year shelf life from date of manufacture

Plum Duo personal eye wash stations are a basic first aid supply for any work environment, and a practical supplement when fixed eye wash stations are present.

Certifications: Meets ANSI Z358.1-2009 (as a personal eye wash only).



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
1000 mL (32 oz.) 0.9% Saline F24880-8103 76169-918 Pack of 6 Retrieving
500 mL (16 oz.) pH Neutralizing F24880-8023 76169-916 Pack of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty