Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plum Duo Covered Eye Wash Stations, Wall Mountable, SP Scienceware

Supplier: Bel-Art Products
Uniquely designed Plum Duo eye wash allows rinsing of both eyes at the same time, eliminating the need to choose which eye to rinse first in an emergency. Plum Duo personal eye wash stations are a basic first aid supply for any work environment, and a practical supplement when fixed eye wash stations are present.

  • Dual eye piece keeps eyes open allowing delivery of solution to both eyes at the same time
  • Two gentle streams of solution flow from the inner eyes toward the outside to keep tear ducts free from further contamination
  • Dispense upright or inverted; quick twist caps break seal for immediate access
  • pH Neutralizing solution neutralizes acids and alkalis to a harmless level in 20 seconds or less
  • 0.9% sterile saline mimics human tears for effective rinsing of dirt and debris
  • Pictogram instructions on stations and bottles overcome language barriers
  • Covered station especially suited for high dust or dirt workspaces
  • Rinse times: pH Neutralizing – approximately 2 min./bottle, Saline – approximately 5 min./bottle
  • No maintenance bottles; 3-year shelf life from date of manufacture

Certifications: Meets ANSI Z358.1-2009 (as a personal eye wash only).

Ordering information: Wall mount hardware included.


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
1000 mL (32 oz.) 0.9% Saline, Two Bottles 30.5 W × 35.6 H cm (12 × 14") F24880-8502 76169-922 Each Retrieving
500 mL (16 oz.) pH Neutralizing and 1000 mL (32 oz.) 0.9% Saline 30.5 W × 35.6 H cm (12 × 14") F24880-8503 76169-924 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty