Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Motic BA410 Elite Hematology Package, Motic Instruments

Supplier: MOTIC INSTRUMENTS INC.
1100100403048
Motic's BA410 Elite Hematology Package is a complete hematology microscopy bundle, which includes everything needed for hematology analysis.

  • Improved interpupillary distance and moveable eyepieces result in maximum comfort for any user
  • Rackless stage can hold two slides at once for quick comparison
  • Absence of gear rack on the stage allows for quick throughput of slides
  • High-end CCIS Achromatic optics minimize chromatic and spherical aberrations
  • A variety of optional upgrades and accessories is availables

This bundle is composed of premium Motic components as well as a wide range of accessories and upgrades making it the perfect choice for any hematologist.

The BA410 Elite Hematology Package takes Motic's flagship life-science microscope (the BA410 Elite) and pairs it with a generous selection of carefully chosen accessories to make it the ultimate hematological microscopy device.

The BA410 Elite's CCIS Plan Achromat Objectives are designed for maximum image fidelity. The expanded resolution power in terms of numerical aperture, along with impeccable color reproduction, yield the most accurate sample representation. The objectives included also provide a high magnification range to give the microscope versatility to handle any hematology task.

Attention has also been paid to ergonomics and comfort. With adjustable eyepieces and rubber eyecups, this microscope can be used for extended periods of time without strain on the eyes or neck.

Further adding to this scope's versatility are its inter changeable halogen and led illumination options. In addition, an ergonomic, tilting head gives the user the comfort needed for extended periods of use.
With all this and more, hematologists need not look any further than Motic's BA410 Elite Hematology Package.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Motic BA410E Hematology Package 1100100403048 10854-398 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Eyepieces Widefield high eyepoint eyepiece WFPL 10×/22 mm with diopter adjustment
Head Siedentopf, 30° Inclined
Condenser Achromat Swing-out condenser N.A.0.90 with iris diaphragm
Nosepiece Sextuple, Reversed
Illumination Koehler illumination quartz halogen 12V/50W and 12V/3W LED module (6000K) with external lamp house and intensity control
Stage Built in low position rackless stage with right hand control

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty