N-Methyl-N-nitrosotoluene-4-sulfonamide ≥97%
|
Danger
|
M23270-25G
101182-216EA
486.78
USD
101182-216
N-Methyl-N-nitrosotoluene-4-sulfonamide ≥97%
N-Methyl-N-nitrosotoluene-4-sulfonamide
Formula:
C₈H₁₀N₂O₃S Melting Pt: 61…62 °C |
UN:
3224 ADR: 4.1,II |
Specification Test Results
Appearance | Light yellow solid |
Storage Temperature | 2 - 8 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...