Modacrylic Jersey Knit ARC Rated Long Sleeve Hi-Vis Shirt, Lakeland Industries
T-FRJ0513L-LG T-FRJ0513L-6X T-FRJ0513L-SM T-FRJ0513L-5X T-FRJ0513L-4X T-FRJ0513L-MD T-FRJ0513L-3X T-FRJ0513L-XL T-FRJ0513L-2X
75923-590EA 131.9 USD
75923-590 75923-736 75923-594 75923-592 75923-586 75923-584 75923-734 75923-732 75923-588
Modacrylic Jersey Knit ARC Rated Long Sleeve Hi-Vis Shirt, Lakeland Industries
Long Sleeve Crew Neck Shirt provides both ANSI 107 Class 3 visibility and CAT 2 ARC flash protection.
  • Crew neck FR/ARC knit shirt
  • Long sleeves
  • Cotton plated inside for comfort
  • 3M Scotchlite reflective material
  • Knit ribbed collar and cuffs
Ideal for Electrical Utility workers and contractors, this Hi-Vis knit combines visibility and arc flash protection in one comfortable garment. Modacrylic fabric will never lose its 10.0 cal/cm2 CAT 2 protective properties and is ANSI 107-Class 3 compliant.
Certifications: ANSI 107-Class 3. ASTM F1506. NFPA 70E, CAT 2.