Vented Back Flame Resistant Shirt, Khaki, Lakeland Industries
NISH7VB20-2X NISH7VB20-SM NISH7VB20-XL NISH7VB20-4X NISH7VB20-MD NISH7VB20-LG NISH7VB20-3X NISH7VB20-5X
75923-572EA 89.9 USD
75923-572 75923-582 75923-570 75923-580 75923-576 75923-574 75923-568 75923-578
Vented Back Flame Resistant Shirt, Khaki, Lakeland Industries
Flame Resistant/ARC rated work shirt with vented back for cooling and increased comfort in hot work environments.
  • Lightweight 7 oz. 88/12 flame resistant material
  • Aramid mesh vented back for cooling and comfort
  • Button down collar
  • Two Chest Pockets
Designed for both comfort and protection, this shirt is dual certified for both flash fire and electrical arc flash protection, all while incorporating an aramid mesh vented back to increase air circulation and keep wearers comfortable in the hottest of work environments. The shirt features a button down collar, long sleeves with button cuffs, two chest pockets, and a long rounded tail for better fit and comfort.
Certifications: NFPA 2112. NFPA 70E, CAT 2.