Aromatase Inhibitor I, Calbiochem®

Supplier: MilliporeSigma

182540-1MG 182540-1MG
80600-270EA 189.02 USD
CA80600-270 80600-270
Aromatase Inhibitor I, Calbiochem®
Aromatase Inhibitor I

A potent, competitive and nonsteroidal inhibitor of aromatase


  • Primary Target: P450 arom
  • White solid form


4-(Imidazolylmethyl)-1-nitro-9H-9-xanthenone


it is more potent than fadrozole in inhibiting aromatase activity. It does not significantly inhibit 17α-hydroxylase.

Formula: C₁₇H₁₁N₃O₄
MW: 321.292 g/mol
Storage Temperature: Freezer
CAS Number: 331684-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR