20X PBS (Phosphate Buffered Saline), 0.1% Tween 20, Teknova
Supplier: TEKNOVA INC
P0291
101446-778EA 82.76 USD
101446-778
20X PBS (Phosphate Buffered Saline), 0.1% Tween 20, Teknova
Buffers Biological Buffers
PBS buffer contains 2.7M sodium chloride, 54mM potassium chloride, 86mM disodium phosphate, 28mM monopotassium phosphate, 0.1% tween-20.