Disposable Surgical Drapes, Stoelting
Supplier: Stoelting
50981 50980
100229-300EA 183.08 USD
100229-300 100229-298
Disposable Surgical Drapes, Stoelting
First Aid Supplies Surgical Drapes
For use in minor surgical or therapeutic procedures where a sterile field or drape is required.
  • Avoids possible moisture or bacteria migration
  • Available in plain or fenestrated
  • Poly interlined, 3-ply construction of tissue/poly/tissue provides superior absorbency