Etidronic acid ≥95%, white powder
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
1-Hydroxyethylidenebis(phosphonic acid)
, 1-Hydroxyethylidenediphosphonic acid
, 1-Hydroxyethylidene-1,1-diphosphonic acid
, HEDP
, 1-Hydroxyethane-1,1-diphosphonic Acid
, OEDFK
, HDEPA
15-2900-5G
15-2900-25G
100199-360EA
294.4
USD
100199-360
100204-954
Etidronic acid ≥95%, white powder
Etidronic acid
Formula:
C₂H₈O₇P₂ MW: 206.028 g/mol |
UN:
3261 ADR: 8,III |
Specification Test Results
Form | Powder |
Color | White |
Odor | Odorless |
Solubility in/Miscibility with water | Insoluble |
Organic solvents | 0% |
VOC content | 0.0 g/l / 0.00 lb/gl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...