The unique Variostat can control the temperature of any size bath from -30 to 150 °C.
- Pilot ONE controller with Plug and Play technology, Precise temperature control, Overtemperature and low level protection, Powerful pressure/suction pump, 5-point calibration, RS232 interface and Pt100 connection
The special construction of the Variostat allows flexibility for the user in a number of different applications. With the infinitely controllable VPC pressure/suction pump, the circulation of the respective bath sizes can be adapted for optimal results. Furthermore, the pump pressure for external applications can be set and controlled using the menu control with an optional pressure sensor. Insulated bath vessels are available in stainless steel as an accessory in three standard sizes or can be custom made with any desired dimensions.