Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Weiler® Hand Tube Brushes

Supplier: ORS NASCO, INC.
Weiler®
804-21090 804-21097 804-21161 804-21095 804-21096 804-21093 804-21094 804-21270 804-21091 804-21092 804-21191 804-21105 804-21139 804-21246 804-21103 804-21233 804-21165 804-21221
Use fully chucked on smooth or threaded blind holes.

Brushes are ideally suited for deburring, cleaning, and finishing applications.



 

ORDER

Expand All / Collapse All
Diameter Overall Length Brush Length Bristle Type Supplier No. VWR Catalog Number Unit Price Quantity
1/2" (1/8" Stem) 8" 2" 0.0060" Steel Wire 804-21097 300049-354 Each Retrieving
1/2" (1/8" Stem) 8" 2" 0.0060" Stainless Steel Wire 804-21105 300049-358 Case of 10 Retrieving
1/2" (1/8" Stem) 8" 2" 0.0040" Brass Wire 804-21165 300049-366 Each Retrieving
1/4" (3/32" Stem) 7" 1 1/2" 0.0030" Steel Wire 804-21092 300050-028 Each Retrieving
1/4" (3/32" Stem) 7" 1 1/2" 0.0050" Steel Wire 804-21093 300050-030 Case of 10 Retrieving
1/4" (3/32" Stem) 7" 1 1/2" 0.0060" Steel Wire 804-21161 300050-038 Case of 10 Retrieving
1/4" (3/32" Stem) 7" 1 1/2" 0.0050" Brass Wire 804-21139 300050-034 Case of 10 Retrieving
1/4" (3/32" Stem) 7" 1 1/2" 0.0030" Brass Wire 804-21270 300050-046 Each Retrieving
3/16" (3/32" Stem) 7" 1 1/2" 0.0050" Steel Wire 804-21091 300050-026 Case of 10 Retrieving
3/16" (3/32" Stem) 7" 1 1/2" 0.0030" Brass Wire 804-21221 300050-042 Case of 10 Retrieving
3/8" (1/8" Stem) 8" 2" 0.0040" Brass Wire 804-21191 300049-368 Each Retrieving
3/8" (1/8" Stem) 8" 2" 0.0060" Stainless Steel Wire 804-21103 300049-356 Case of 10 Retrieving
3/8" (1/8" Stem) 8" 2" 0.0060" Steel Wire 804-21095 300049-350 Each Retrieving
3/8" (1/8" Stem) 8" 2" 0.0040" Steel Wire 804-21094 300049-348 Each Retrieving
9/16" (1/8" Stem) 9" 2 1/2" 0.0050" Steel Wire 804-21233 300049-372 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty