Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Weiler® Double-Spiral, Double-Stem Power Tube Brushes

Supplier: ORS NASCO, INC.
Weiler®
804-21252 804-21184 804-21106 804-21117 804-21114 804-21115 804-21179 804-21113 804-21124 804-21110 804-21111 804-21232 804-21109 804-21107 804-21108
Use fully chucked on smooth or threaded blind holes.

Brushes are ideally suited for deburring, cleaning, and finishing applications. For use with power tools.



 

ORDER

Expand All / Collapse All
Diameter Overall Length Brush Length Bristle Type Supplier No. VWR Catalog Number Unit Price Quantity
1/2" (5/32" Stem) 5" 2" 0.0060" Steel Wire 804-21107 300010-200 Case of 10 Retrieving
1/2" (5/32" Stem) 5" 2" 0.0040" Steel Wire 804-21106 300049-360 Each Retrieving
1/2" (5/32" Stem) 5" 2" 0.0104" Steel Wire 804-21252 300049-374 Case of 10 Retrieving
1/2" (5/32" Stem) 5" 2" 0.0060" Stainless Steel Wire 804-21117 300049-362 Case of 10 Retrieving
3/4" (1/4" Stem) 5 1/2" 2 1/2" 0.0104" Steel Wire 804-21111 300048-598 Case of 10 Retrieving
3/4" (1/4" Stem) 5" 2 1/2" 0.0060" Brass Wire 804-21179 300048-600 Case of 10 Retrieving
3/4" (1/4" Stem) 5" 2 1/2" 0.0060" Steel Wire 804-21110 300010-201 Each Retrieving
3/8" (1/8" Stem) 6" 2 1/8" 0.0060" Steel Wire 804-21232 300049-370 Case of 10 Retrieving
5/8" (7/32" Stem) 5" 2" 0.0080" Steel Wire 804-21109 300048-958 Each Retrieving
5/8" (7/32" Stem) 5" 2" 0.0104" Steel Wire 804-21184 300048-966 Case of 10 Retrieving
5/8" (7/32" Stem) 5" 2" 0.0050" Steel Wire 804-21108 300048-956 Each Retrieving
7/8" (1/4" Stem) 5 1/2" 2 1/2" 0.0104" Steel Wire 804-21113 300048-462 Each Retrieving
1" (1/4" Stem) 5 1/2" 2 1/2" 0.0104" Steel Wire 804-21115 300010-202 Each Retrieving
1" (1/4" Stem) 5 1/2" 2 1/2" 0.0060" Steel Wire 804-21114 300048-464 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty