3,5-Dibromoaniline 98%

Warning

AC46056-0050 AC46056-0250
103465-112EA 91.8 USD
103465-112 103465-114
3,5-Dibromoaniline 98%
3,5-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 53…54.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00047841
CAS Number: 626-40-4

Order Now

Specification Test Results

Appearance Brown to black Solid or crystalline solid
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR