Methyl oleate ≥99% analytical standard for GC

Supplier: ACROS ORGANICS MS

Synonyms: Oleic acid methyl ester

AC46038-0050 AC46038-0010
103465-902EA 84.86 USD InStock
103465-902 103465-900
Methyl oleate ≥99% analytical standard for GC
Methyl oleate
Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 186°C (9 mm Hg)
Melting Pt: –20 °C
Density: 0.87
Flash Pt: 180°C
Storage Temperature: Refrigerator
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 12,06965

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4530 (20°C, 589 nm)
GC ≥98.5 %
Appearance Clear colorless Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR