Methyl myristate 99% analytical standard for GC

Synonyms: Myristic acid methyl ester Tetradecanoic acid methyl ester Tetradecylic acid methyl ester Methyl tetradecanoate

AC46036-0050 AC46036-0010
103465-892EA 42 USD
103465-892 103465-894
Methyl myristate 99% analytical standard for GC
Methyl myristate
Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323 °C (760 mmHg)
Melting Pt: 18.4 °C
Density: 0.863 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008983
CAS Number: 124-10-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR