Methyl octanoate 99% analytical standard for GC

Synonyms: Caprylic acid methyl ester Octanoic acid methyl ester Methyl caprylate

AC46035-0010 AC46035-0050
103465-898EA 89.89 USD
103465-898 103465-896
Methyl octanoate 99% analytical standard for GC
Methyl octanoate
Formula: C₉H₁₈O₂
MW: 158.24 g/mol
Boiling Pt: 79 °C (12 mmHg)
Density: 0.878 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00009551
CAS Number: 111-11-5

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4185 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR