2-Bromo-1-chloro-4-iodobenzene 98%

Warning

AC45838-0050 AC45838-0010
103464-276EA 280 USD
103464-276 103464-274
2-Bromo-1-chloro-4-iodobenzene 98%
2-Bromo-1-chloro-4-iodobenzene
Formula: C₆H₃BrCl
MW: 317.3495 g/mol
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD07780667
CAS Number: 31928-46-8

Order Now

Specification Test Results

Appearance Yellow to brown Powder or crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 39 to 44 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR