2-Bromopyrazine ≥95%

Supplier: ACROS ORGANICS MS

Danger

AC45816-0010 AC45816-0050
103464-408EA 89.85 USD
103464-408 103464-410
2-Bromopyrazine ≥95%
2-Bromopyrazine
Formula: C₄H₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD08275680
CAS Number: 56423-63-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Appearance Clear colorless to light yellow Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR