2-Bromoquinoline 98%

Warning

AC45764-0250
103464-418EA 872.7 USD
103464-418
2-Bromoquinoline 98%
2-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00234480
CAS Number: 2005-43-8

Order Now

Specification Test Results

Appearance White to yellow to red Powder
Infrared spectrum Conforms
Melting point 45 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR