2-Chloro-4-formylpyridine 97%

Warning

Synonyms: 2-Chloroisonicotinaldehyde2-Chloropyridine-4-carboxaldehyde

AC45604-0050 AC45604-0010
103464-536EA 310.3 USD
103464-536 103464-534
2-Chloro-4-formylpyridine 97%
2-Chloro-4-formylpyridine
Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 46…47 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD06651557
CAS Number: 101066-61-9

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 44 to 50 °C
Water ≤1.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR