N-Methyl-2,2,2-trifluoroacetamide

Warning

Synonyms: N-Methyltrifluoroacetamide2,2,2-Trifluoro-N-methylacetamide

AC22157-0050 AC22157-0250
103466-162EA 181.9 USD
103466-162 103466-160
N-Methyl-2,2,2-trifluoroacetamide
N-Methyl-2,2,2-trifluoroacetamide
Formula: C₃H₄F₃NO
MW: 127.07 g/mol
Boiling Pt: 156…157 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00009670
CAS Number: 815-06-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 52 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR