Chromotrope 2B, pure

Supplier: ACROS ORGANICS MS

Warning

AC14975-0250 AC14975-0050
103463-790EA 54.4 USD
103463-790 103463-788
Chromotrope 2B, pure
Chromotrope 2B
Formula: C₁₆H₉N₃Na₂O₁₀S₂
MW: 513.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003951
CAS Number: 548-80-1
EINECS: 208-959-9

Order Now

Specification Test Results

Appearance Dark green to greenish-brown to brown Powder
Infrared spectrum Conforms
Loss on drying ≤18 %
UV Lambda max 513 to 517 nm (c=0.1, H2O)
Specific extinction E (1%/1cm) ≥580 (on dry substance)
Appearance Dark green to greenish-brown to brown Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR