Salicylaldehyde azine 99%

Warning

Synonyms: SalicylaldazineSalazineBis(2-hydroxybenzaldehyde) hydrazoneN,N'-Bis(salicylidene)hydrazine

AC45694-0050 AC45694-0250 AC45694-1000
103466-452EA 608 USD
103466-452 103466-450 103466-448
Salicylaldehyde azine 99%
Salicylaldehyde azine
Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00043496
CAS Number: 959-36-4

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR