Would you like to log in again?
BioFit® Combination Clean Room/ESD Chairs and Stools, BioFit
Supplier: BIOFIT ENGINEERED SEATING
ETALR1000KFFML ETSLR-C10KMBLU BTSMH-C1-K-MBL ETSLR1000KMBLU EESMHVUV-KMBLU EESH-HC1K-MBLU EESH-HVUVKMBLU BTSMHG1000KMBL BTSHRVUVKMBLU EEAMHVUV-KFMBL BTSHHGC10KMBLU EEAMHC1KATFMBL ETAMHC1KFASMBL ETSHHVUVK-MBLU ETAMHC10KFASMU EESLRC10K-MBLU BTSHH1000KMBLU ETAMHC1KATFMBL EESMH-C10KMBLU BTSMH-VUVK-MBL ETSHH-C1K-MBLU EEAMHC10K-MBLU ETAMH-VUVKMBLU EEAMH-VUV-KMBL EEWMHVUVKFMBLU EEALR-VUVKMBL EEALR1000K-MBL ETAMH1000KFMBL ETALRC1KASMBLU EEAMHC10KFMBLU ETSHHC10KMBLU EESH-HC10KMBLU EESLR1000KMBLU BTSMR-VUVK-MBL ETAMH-C1K-MBLU ETSLR-C1K-MBLU ETALR-C1K-MBLU ETSHH1000KMBLU BTSLR-VUVK-MBL ETSMH-C10KMBLU EEAMH1000KFMBL BTSLR1000KMBLU BTSLRC10K-MBLU ETAMHC10KFMBLU ETAMHVUVKFMBLU ETAMHC10K-MBLU EESMH-C1K-MBLU ETALRC10KASMBL ETSMH-C1K-MBLU EESH-H1000KMBL ETALRVUVKFFMBL EEALR-C10K-MBL ETAMH1000KMBLU EESLRVUV-KMBLU ETAMH1000KFFFU ETSLR-VUVKMBLU ETALRVUVK-MBLU EESMH1000KMBLU BTSLRC1-KMBLU ETSMH-VUVKMBLU EEWLRVUV-KMBLU ETALRC10K-MBLU EEALR-C1K-MBLU EEWMHVUVK-MBLU ETALR1000KMBLU BTSHHGVUVKMBLU EEWLHVUV-KMBLU BTSMHC10K-MBLU EESLR-C1K-MBLU EEAMH-C1K-MBLU ETSMH1000KMBLU EESMR1000KMBLU BTSHHGC1-KMBLU EEAMH-1000KMBL ETAMHVUVKFFFMU
75821-156EA 1151.72 USD InStock
75821-156 75821-258 75796-786 75821-254 75821-390 75821-102 75821-112 75796-776 75796-740 75821-084 75796-714 75821-068 75821-186 75821-246 75821-500 75821-386 75796-708 75821-188 75821-134 75796-798 75821-236 75821-076 75821-216 75821-086 75821-416 75796-876 75796-856 75821-490 75821-422 75796-888 75821-232 75821-098 75821-382 75796-810 75821-190 75821-262 75821-424 75821-226 75796-768 75821-272 75796-880 75796-750 75796-756 75821-502 75821-214 75821-178 75821-138 75821-164 75821-276 75821-376 75821-434 75796-860 75821-492 75821-126 75821-488 75821-450 75821-436 75821-130 75796-760 75821-284 75821-406 75821-166 75796-864 75821-418 75821-158 75796-728 75821-400 75796-782 75821-116 75821-070 75821-268 75821-396 75796-718 75796-882 75821-212
BioFit® Combination Clean Room/ESD Chairs and Stools, BioFit
Chairs
BioFit’s Clean Room/ESD chairs are designed and tested to meet ISO 14644-1 cleanliness standards and still provide user comfort, health and wellbeing.
  • Microcon blue vinyl upholstery
  • Backrest height and tilt adjustment
  • Spring-loaded contact ball bearing assembly assures continuity thoughout the chair
  • Drag wire for added continuity from the chair to the ground
  • Chrome-plated metal parts
  • Ergonomic comfort and support
  • Dual-wheel resistance clean room/ESD casters enhance user safety by inhibiting seating from rolling away on hard floors or 2" high polyamide glides
  • Adjustable and fixed height footrings
BioFit’s seating solutions are crafted with user comfort, health and wellbeing in mind
BioFit has specialized in the development of seating for technical applications since our founding, offering more solutions for combination Clean Room/ESD environments than any other manufacturer.
Over 70 years’ background improving the workplace experience to provide ergonomic, LEED® compliant seating for healthcare, technology, education, lab, industrial, office and custom applications.
Products meet or exceed rigorous industry standard and carry a 13-year warranty with a lifetime warranty on the pneumatic mechanisms.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty