    Sort by:
  • Search Within Results
MAL-dPEG®12-NHS ester
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  QUANTA BIODESIGN MS
Description:   Single compound - dPEG® Spacer is 46 atoms and 53.3 Å