    Sort by:
You Searched For:
Bis-MAL-Lysine-dPEG®4-acid
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  QUANTA BIODESIGN MS
Description:   Single compound - dPEG® Spacer is atoms and 17.5 and 27.9 Å, resp.