    Sort by:
You Searched For:
Bis-dPEG®17-PFP ester
1  results were found
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  QUANTA BIODESIGN MS
Description:   Single compound - dPEG® Spacer is 55 atoms and 64.4 Å