    Sort by:
  • Search Within Results
Bis-dPEG®13-NHS ester
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  QUANTA BIODESIGN MS
Description:   Single compound - dPEG® Spacer is 43 atoms and 50.1 Å