    Sort by:
  • Search Within Results
Azido-dPEG®7-amine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  QUANTA BIODESIGN MS
Description:   Single compound - dPEG® Spacer is 25 atoms and 28.8 Å