You Searched For: Amino-PEG4-acid
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Ambeed
Description: 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid, Purity: 95%, CAS number: 663921-15-1, Appearance: Colorless or White to Yellow Solid or liquid, Storage: Sealed in dry, 2-8C, Size: 5G