Weiler® Tiger Abrasives Adapting Nuts, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-56494
300010-291PK 49.11 USD
300010-291
Weiler® Tiger Abrasives Adapting Nuts, ORS Nasco
Fasteners Nuts
Adapting nuts for use with Tiger, Vortec Pro, and high density flap discs.
  • Can be mounted with threaded hole
  • Use with right angle air die grinder or cut off die grinder
  • 5/8" - 11 Arbor thread