1-Vinyl-2-pyrrolidone stabilized
Supplier: Pfaltz & Bauer
|
Danger
|
Synonyms:
N-Vinyl-2-pyrrolidinone
, N-Vinyl-2-pyrrolidone
V02450-100G
101184-018EA
227.94
USD
101184-018
1-Vinyl-2-pyrrolidone stabilized
1-Vinyl-2-pyrrolidone
Formula:
C₆H₉NO MW: 111.1418 g/mol Boiling Pt: 92…95 °C Melting Pt: 13 °C Density: 1.04 Flash Pt: 95 °C |
MDL Number:
MFCD00003197 CAS Number: 88-12-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...