DBCO-TFP ESTER

11815 11815
103575-502EA 323 USD
103575-502 103389-436
DBCO-TFP ESTER
DBCO-TFP ESTER
Formula: C₂₆H₁₇F₁₄NO₃
MW: 467.41 g/mol
CAS Number: 2247993-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR