SP Wilmad-LabGlass Variable Speed Air Stirrer, SP Industries
Supplier: SP Industries
LG-9574-100
14009-540EA 862.58 USD
14009-540
SP Wilmad-LabGlass Variable Speed Air Stirrer, SP Industries
This variable speed air stirrer has a stainless steel shaft (3/8 " dia.) and propeller.
  • It is ideal for stirring organic solvents, paints, etc. where danger of explosion exists
  • Speed is controlled by an air supply valve
  • Supplied with a muffler for quiet operation
Featuring gear reduction, fluctuations in speed due to changes in air pressure are kept to a minimum.