CONSTIX®, Sealed Polyester Swabs, SP-10M Series, Contec®
Supplier: Contec
CONSTIX®
SP-10M
76057-702CS 91.61 USD
76057-702
CONSTIX®, Sealed Polyester Swabs, SP-10M Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-10M sealed microfiber swabs consist of 100% polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Thermally sealed swab is compatible with most common solvents, such as alcohols and ketones
  • Flexible tip enables access to very small areas and component
The cleanroom laundered, double-layer knitted polyester head provides superior sorbency, particle entrapment, and residue removal. The head is thermally sealed and attached, constructed without adhesives or binders. The polypropylene handle has a thin, flexible tip. The swab is durable, absorbent, and features low particles and residue levels.