CONSTIX®, Sealed Polyester Swabs, SP-10 Series, Contec®
Supplier: Contec
CONSTIX®
SP-10
76057-700CS 77.11 USD
76057-700
CONSTIX®, Sealed Polyester Swabs, SP-10 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-10 sealed polyester swabs consist of 100% knitted polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Polyester head provides enhanced sorbency, particle entrapment, and removal
  • Polyester head is compatible with most common solvents, such as alcohols and ketones
The swab head is made of cleanroom laundered, double-layer knitted polyester material. The polyester head is thermally sealed and attached, constructed without adhesives or binders. The polypropylene handle has a thin, flexible tip that enables access to very small areas and components. The swab is durable, absorbent, and features extremely low particles and residue levels.