Polyethylene glycol 600 ≥100% (v/v)
Supplier: Rigaku Reagents
Synonyms:
PEG 600
1008416
1008417
101443-482EA
77.49
USD
101443-482
101443-872
Polyethylene glycol 600 ≥100% (v/v)
Polyethylene glycol 600
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Formula:
H(OCH₂CH₂)nOH MW: 600 g/mol |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...