Magnesium formate 0.5 M

Supplier: Rigaku Reagents

1008044 1008045
101443-372EA 85 USD
101443-372 101443-730
Magnesium formate 0.5 M
Magnesium formate solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: C₂H₂MgO₄·2H₂O
MDL Number: MFCD00150512
CAS Number: 6150-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR