Magnesium chloride 1 M

Supplier: Rigaku Reagents

Synonyms: D-Gulonic acid γ-lactoneD-Gulonic acid-1,4-lactone

1008237 1008236
101443-370EA 75.85 USD
101443-370 101443-726
Magnesium chloride 1 M
Magnesium chloride solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: MgCl₂·6H₂O
MDL Number: MFCD00149781
CAS Number: 7791-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR