Ammonium sulfite 1 M

Supplier: Rigaku Reagents

1008110 1008109
101443-838EA 91.54 USD
101443-838 101443-446
Ammonium sulfite 1 M
Ammonium sulphite solutions

Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.

Formula: H₈N₂O₃S·1H₂O
MDL Number: MFCD00066094
CAS Number: 7783-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR