SP Wilmad-LabGlass Stir Bar Retrievers, SP Industries
Supplier: SP Industries
LG-9569T-100 LG-9569T-200 LG-9569T-110
14201-282EA 21.34 USD
14201-282 14212-554 14201-266
SP Wilmad-LabGlass Stir Bar Retrievers, SP Industries
Retrieve stir bars without messy spills or breakage.
  • PTFE or polyethylene casing available
A 3/8" Alnico V magnet is completely enclosed in a PTFE or polyethylene casing and attached to rod of the same material. PTFE retriever features a flexible shaft , for hard-to-reach stir bars, whereas the polyethylene version is rigid with hanging loop.