Celestron Microspin 2MP Digital Microscope
44114
470223-718EA 131.3 USD
470223-718
Celestron Microspin 2MP Digital Microscope
Six objectives in vertically mounted objective wheel
  • 100x, 200x, 300x, 400x, 500x, 600x magnification
  • MicroSpin Capture Pro Software DVD/CD included
  • Windows and Apple OS compatible
The intermediate-level MicroSpin is a 2 MP desktop digital microscope with a six objectives wheel. A 2 MP imaging sensor paired with upper and lower illumination for reflective and transparent illumination provides excellent resolution and illumination so you can capture high quality still images and video. A simple, non-adjustable stage with plastic clips holds slides in place for an optimized viewing experience.
The objective wheel rotates clockwise and counterclockwise, and locates in place at each position, making it easy to change powers. With six magnification powers from 100x up to 600x, this microscope is perfect for students and professionals alike.