SP Wilmad-LabGlass Indented Cylindrical Reaction Vessels, O-Ring Flange, SP Industries
Supplier: SP Industries
LG-8079A-104 LG-8079A-106 LG-8079A-102 LG-8079A-100
14204-366EA 344.68 USD
14204-366 14209-310 14204-838 14207-840
SP Wilmad-LabGlass Indented Cylindrical Reaction Vessels, O-Ring Flange, SP Industries
These borosilicate glass reaction vessels have four indentations for improved stirring, and a tooled flat flange with an O-ring groove for a greaseless seal.