Reagents for Coulometric Karl Fischer Titration, Thermo Scientific
Supplier: Thermo Scientific
47117-RDL 47117-PDL
AA47117-PDLEA 109.35 USD
AA47117-PDL AA47117-RDL
Reagents for Coulometric Karl Fischer Titration, Thermo Scientific
Karl Fischer Titrants and Reagents
Coulometric titration can be carried out in two types of titration cells - with or without a diaphragm
Cells with a diaphragm were developed first and used a semi-permeable glass diaphragm to separate the volume of the cell into two chambers containing the cathode and the anode, respectively. The separation was used to isolate the individual electrode processes and avoid reduction of iodine back to iodide.