SP Wilmad-LabGlass Half-Jacketed Spherical Reaction Vessels with 28/15 Ball Joint, Conical Flange, SP Industries
Supplier: SP Industries
LG-8026-104 LG-8026-102
14207-526EA 2785 USD
14207-526 14206-144
SP Wilmad-LabGlass Half-Jacketed Spherical Reaction Vessels with 28/15 Ball Joint, Conical Flange, SP Industries
Made of borosilicate glass, these spherical reaction vessels have a hemispherical half-jacket and are equipped with 1/2" hose connectors, for either heating or cooling applications.
Bottom of vessel has a [SJ] 28/15 ball joint for connection to LG-8027 or LG-8028T take-off adapter
NOTE: LG-8070-100 clamp is used with these flasks.
Top is finely ground to provide a leak-proof fit with lids LG-8030 to LG-8050.