    Sort by:
  • Search Within Results
Sepabeads® SP825L
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Alfa Aesar
Description:   Sepabeads SP825L is used in amino acid separations, small molecules in medium hydrophobicity in polar solvents.