You Searched For: Sepabeads® SP710
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Catalog Number: (AA46555-A1)
Supplier: Thermo Scientific Chemicals
Description: Sepabeads* SP710, synthetic adsorbent resin, highly porous type, PS-DVB, P.R. 90 angstroms, Form: 250um Beads, S.A. 900m2/g, Size: 1KG